Assay Details
Gene Symbol: Atic | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031014.2 | NP_112276.2 | 10 - 11 | 1074 | 85 | |
GenBank mRNA | BC072496.1 | - | 10 - 11 | 1074 | 85 | |
D89514.1 | - | 10 - 11 | 1067 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
