Assay Details
Gene Symbol: Pecr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133299.2 | NP_579833.1 | 2 - 3 | 355 | 98 | |
XM_008767180.1 | XP_008765402.1 | 2 - 3 | 361 | 98 | ||
GenBank mRNA | AF021854.1 | - | 2 - 3 | 356 | 98 | |
AF099742.1 | - | 2 - 3 | 275 | 98 | ||
BC060546.1 | - | 2 - 3 | 349 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
