Assay Details
Gene Symbol: Pecr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133299.2 | NP_579833.1 | 3 - 4 | 513 | 64 | |
XM_008767180.1 | XP_008765402.1 | 3 - 4 | 519 | 64 | ||
GenBank mRNA | AF021854.1 | - | 3 - 4 | 514 | 64 | |
AF099742.1 | - | 3 - 4 | 433 | 64 | ||
BC060546.1 | - | 3 - 4 | 507 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
