Assay Details
Gene Symbol: Acadl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012819.1 | NP_036951.1 | 4 - 5 | 546 | 74 | |
GenBank mRNA | BC062006.1 | - | 4 - 5 | 544 | 74 | |
FQ215575.1 | - | 4 - 5 | 597 | 74 | ||
J05029.1 | - | 4 - 5 | 557 | 74 | ||
L11276.1 | - | 4 - 5 | 546 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
