Assay Details
Gene Symbol: Acadl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012819.1 | NP_036951.1 | 6 - 7 | 776 | 131 | |
GenBank mRNA | BC062006.1 | - | 6 - 7 | 774 | 5599828 | 131 |
J05029.1 | - | 6 - 7 | 787 | 131 | ||
L11276.1 | - | 6 - 7 | 776 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
