Assay Details
Gene Symbol: Acadl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012819.1 | NP_036951.1 | 8 - 9 | 987 | 116 | |
GenBank mRNA | BC062006.1 | - | 8 - 9 | 985 | 116 | |
J05029.1 | - | 8 - 9 | 998 | 116 | ||
L11276.1 | - | 8 - 9 | 987 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
