Assay Details
Gene Symbol: Bard1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022622.1 | NP_072144.1 | 7 - 8 | 1666 | 81 | |
GenBank mRNA | AF182946.1 | - | 7 - 8 | 1666 | 81 | |
AY583210.1 | - | 4 - 5 | 1360 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
