Assay Details
Gene Symbol: Rpe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006245104.2 | XP_006245166.1 | 5 - 6 | 670 | 61 | |
XM_006245105.2 | XP_006245167.1 | 5 - 6 | 652 | 61 | ||
XM_006245106.2 | XP_006245168.1 | 5 - 6 | 633 | 61 | ||
XM_006245107.2 | XP_006245169.1 | 4 - 5 | 592 | 61 | ||
XM_006245109.2 | XP_006245171.1 | 4 - 5 | 573 | 61 | ||
XM_008767287.1 | XP_008765509.1 | 5 - 6 | 689 | 61 | ||
XM_008767289.1 | XP_008765511.1 | 5 - 6 | 544 | 61 | ||
XM_008767290.1 | XP_008765512.1 | 4 - 5 | 557 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
