Assay Details
Gene Symbol: Mzf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006228116.2 | XP_006228178.1 | 1 - 2 | 320 | 61 | |
XM_008758768.1 | XP_008756990.1 | 1 - 2 | 320 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006228116.2, XM_008758768.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
