Assay Details
Gene Symbol: Il17rd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191937.1 | NP_001178866.1 | 12 | 1985 | 61 | |
XM_006252636.2 | XP_006252698.1 | 12 | 2115 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
