Assay Details
Gene Symbol: Arhgef3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106061.1 | NP_001099531.1 | 6 - 7 | 730 | 66 | |
XM_006252574.2 | XP_006252636.1 | 9 - 10 | 862 | 66 | ||
XM_006252575.2 | XP_006252637.1 | 9 - 10 | 1060 | 66 | ||
XM_006252576.2 | XP_006252638.1 | 7 - 8 | 1226 | 66 | ||
XM_008770982.1 | XP_008769204.1 | 9 - 10 | 927 | 66 | ||
XM_008770983.1 | XP_008769205.1 | 9 - 10 | 769 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
