Assay Details
Gene Symbol: Rfc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009629.1 | NP_001009629.1 | 4 - 5 | 451 | 71 | |
XM_006248734.2 | XP_006248796.1 | 3 - 4 | 342 | 71 | ||
GenBank mRNA | BC088281.1 | - | 4 - 5 | 451 | 7383278 | 71 |
FQ216316.1 | - | 4 - 5 | 440 | 71 | ||
FQ232102.1 | - | 4 - 5 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
