Assay Details
Gene Symbol: Cadm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047102.1 | NP_001040567.1 | 3 - 4 | 505 | 83 | |
GenBank mRNA | DQ272744.1 | - | 3 - 4 | 505 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
