Assay Details
Gene Symbol: Ttc33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106414.1 | NP_001099884.1 | 2 - 3 | 416 | 82 | |
XM_006231993.2 | XP_006232055.1 | 2 - 3 | 305 | 82 | ||
XM_006231994.1 | XP_006232056.1 | 2 - 3 | 286 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
