Assay Details
Gene Symbol: Il7r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106418.1 | NP_001099888.1 | 3 - 4 | 449 | 91 | |
XM_006257698.2 | XP_006257760.1 | 3 - 4 | 333 | 91 | ||
XM_006257699.2 | XP_006257761.1 | 3 - 4 | 307 | 91 | ||
XM_008773706.1 | XP_008771928.1 | 2 - 3 | 2245 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
cell morphogenesis
negative regulation of T cell mediated cytotoxicity
immunoglobulin production
biological_process
regulation of cell size
positive regulation of gene expression
cell growth
cytokine-mediated signaling pathway
T cell differentiation
positive regulation of T cell differentiation in thymus
B cell proliferation
lymph node development
homeostasis of number of cells
molecular_function
cytokine receptor activity
cell morphogenesis
negative regulation of T cell mediated cytotoxicity
immunoglobulin production
biological_process
regulation of cell size
positive regulation of gene expression
cell growth
cytokine-mediated signaling pathway
T cell differentiation
positive regulation of T cell differentiation in thymus
B cell proliferation
lymph node development
homeostasis of number of cells
molecular_function
cytokine receptor activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P007943 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P828142 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 665416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 707518 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 567903 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53208 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187853 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330009 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553685 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83505 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650777 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 88576 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 86739 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 950774 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405228 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399385 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102647 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742250 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 282485 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123776 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 466699 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94581 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 3110 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40413 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P828142 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P007943 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P828142 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P007943 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P828142 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P007943 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P100584 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 87554 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10821852 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10821853 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10821858 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10821855 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10821857 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10821859 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10821854 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10821856 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10821851 | - | - |
Rat Genome 230 2.0 Array | 1382566_PM_at | - | - |
Rat Genome 230 2.0 Array | 1386052_PM_at | - | - |
Rat Genome 230 2.0 Array | 1386052_at | - | - |
Rat Genome 230 2.0 Array | 1382566_at | - | - |
Affymetrix HT Rat Focus Array | 1382566_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1386052_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1386052_at | - | - |
Affymetrix HT Rat Focus Array | 1382566_at | - | - |
Affymetrix Rat Clariom S Assay | PSR2_KL567908v1_random00000006.rn.1 | TC2_KL567908v1_random00000002.rn.1 | RTA TC |
Rat Expression Set 230 | 1386052_PM_at | - | - |
Rat Expression Set 230 | 1386052_at | - | - |
Rat Expression Set 230 | 1382566_at | - | - |
Rat Expression Set 230 | 1382566_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10821851 | - | - |
Rat Genome U34 Set | rc_AA800754_at | - | - |
Rat Genome U34 Set | rc_AI011994_at | - | - |
Rat Genome U34 Set | rc_AI071569_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21993897 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1358648 | - | - |
