Assay Details
Gene Symbol: Skp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106416.2 | NP_001099886.1 | 5 - 6 | 758 | 64 | |
GenBank mRNA | BC166459.1 | - | 5 - 6 | 758 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
