Assay Details
Gene Symbol: Nadk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044252.1 | NP_001037717.1 | 12 - 13 | 1131 | 85 | |
XM_006232014.2 | XP_006232076.1 | 12 - 13 | 1425 | 85 | ||
XM_006232015.2 | XP_006232077.1 | 11 - 12 | 1358 | 85 | ||
XM_006232016.2 | XP_006232078.1 | 13 - 14 | 1211 | 85 | ||
GenBank mRNA | BC105901.1 | - | 8 - 9 | 715 | 7375971 | 85 |
DQ504300.1 | - | 12 - 13 | 1131 | 85 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P22136 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 473546 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406484 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 515488 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668114 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683776 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396526 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 822986 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514260 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 292905 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418289 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 303401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479668 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 631745 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 876294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333204 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866613 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 740496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94809 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422728 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786674 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726705 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115877 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444835 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900027 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184268 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 367402 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294222 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380274 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 426338 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 279103 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 780581 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 107020 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125567 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 127505 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71828 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33695 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P22136 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P22136 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P22136 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P633413 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P22136 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P354824 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 4235 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 83791 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 36222 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10813585 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813581 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813579 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813586 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813593 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813580 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813589 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813584 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813591 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813590 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813592 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813583 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813582 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10813578 | - | - |
Rat Genome 230 2.0 Array | 1382235_PM_at | - | - |
Rat Genome 230 2.0 Array | 1382235_at | - | - |
Affymetrix HT Rat Focus Array | 1382235_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1382235_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0200003378.rn.1 | TC0200000540.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0200003380.rn.1 | TC0200000540.rn.1 | RTA TC |
Rat Expression Set 230 | 1382235_PM_at | - | - |
Rat Expression Set 230 | 1382235_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10813578 | - | - |
Rat Genome U34 Set | rc_AI233081_at | - | - |
Rat Genome U34 Set | rc_AA818258_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 20995827 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1357630 | - | - |
