Assay Details
Gene Symbol: Nadk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044252.1 | NP_001037717.1 | 2 - 3 | 251 | 68 | |
XM_006232014.2 | XP_006232076.1 | 1 - 2 | 479 | 68 | ||
XM_006232015.2 | XP_006232077.1 | 1 - 2 | 478 | 68 | ||
GenBank mRNA | DQ504300.1 | - | 2 - 3 | 251 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
