Assay Details
Gene Symbol: Nadk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044252.1 | NP_001037717.1 | 9 - 10 | 903 | 69 | |
XM_006232014.2 | XP_006232076.1 | 9 - 10 | 1197 | 69 | ||
XM_006232015.2 | XP_006232077.1 | 8 - 9 | 1130 | 69 | ||
XM_006232016.2 | XP_006232078.1 | 10 - 11 | 983 | 69 | ||
GenBank mRNA | BC105901.1 | - | 5 - 6 | 487 | 69 | |
DQ504300.1 | - | 9 - 10 | 903 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
