Assay Details
Gene Symbol: Lpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012598.2 | NP_036730.1 | 6 - 7 | 1202 | 92 | |
GenBank mRNA | BC081836.1 | - | 6 - 7 | 1202 | 7106121 | 92 |
L03294.1 | - | 1187 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
