Assay Details
Gene Symbol: Ccl25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037203.1 | NP_001032280.1 | 2 - 3 | 165 | 74 | |
XM_006248766.2 | XP_006248828.1 | 2 - 3 | 275 | 74 | ||
GenBank mRNA | BC107945.1 | - | 2 - 3 | 165 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
