Assay Details
Gene Symbol: Ccl25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037203.1 | NP_001032280.1 | 4 - 5 | 402 | 120 | |
XM_006248766.2 | XP_006248828.1 | 4 - 5 | 512 | 120 | ||
GenBank mRNA | BC107945.1 | - | 4 - 5 | 402 | 7388091 | 120 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
