Assay Details
Gene Symbol: Ifrd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047871.1 | NP_001041336.1 | 9 - 10 | 1242 | 70 | |
GenBank mRNA | BC161810.1 | - | 9 - 10 | 1176 | 70 | |
DQ480751.1 | - | 9 - 10 | 1242 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
