Assay Details
Gene Symbol: Rab6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108775.1 | NP_001102245.1 | 6 - 7 | 491 | 64 | |
XM_006243669.2 | XP_006243731.1 | 7 - 8 | 795 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
retrograde vesicle-mediated transport, Golgi to endoplasmic reticulum
intra-Golgi vesicle-mediated transport
metabolic process
Rab protein signal transduction
retrograde transport, endosome to Golgi
GTPase activity
GTP binding
GDP binding
myosin V binding
intracellular protein transport
retrograde vesicle-mediated transport, Golgi to endoplasmic reticulum
intra-Golgi vesicle-mediated transport
metabolic process
Rab protein signal transduction
retrograde transport, endosome to Golgi
GTPase activity
GTP binding
GDP binding
myosin V binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P031855 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P569832 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517932 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863239 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389510 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112599 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 857656 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174356 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 705904 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 501038 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 414604 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588177 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966227 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 840490 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 415151 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240058 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 148309 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519963 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332502 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207896 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 615350 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947140 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275456 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640948 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 272493 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 176385 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580525 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173364 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 772772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 808345 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 139034 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31519 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 47527 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43627 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167357 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175137 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P569832 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P031855 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P569832 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P031855 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P031855 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P569832 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P291938 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P321583 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P569832 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 62534 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 55546 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 88305 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10912638 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912633 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912637 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912635 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912636 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912634 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912639 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10912631 | - | - |
Rat Genome 230 2.0 Array | 1392478_PM_at | - | - |
Rat Genome 230 2.0 Array | 1388632_at | - | - |
Rat Genome 230 2.0 Array | 1388632_PM_at | - | - |
Rat Genome 230 2.0 Array | 1392478_at | - | - |
Affymetrix HT Rat Focus Array | 1392478_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388632_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388632_at | - | - |
Affymetrix HT Rat Focus Array | 1392478_at | - | - |
Rat Expression Set 230 | 1388632_PM_at | - | - |
Rat Expression Set 230 | 1388632_at | - | - |
Rat Expression Set 230 | 1392478_at | - | - |
Rat Expression Set 230 | 1392478_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10912631 | - | - |
Rat Genome U34 Set | rc_AI007905_at | - | - |
Rat Genome U34 Set | rc_AI009969_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22165389 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353058 | - | - |
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1530445 | - | - |
