Assay Details
Gene Symbol: Osbpl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006244025.2 | XP_006244087.1 | 10 - 11 | 2126 | 93 | |
XM_008766657.1 | XP_008764879.1 | 10 - 11 | 2183 | 93 | ||
XM_243980.7 | XP_243980.4 | 10 - 11 | 3278 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
