Assay Details
Gene Symbol: Osbpl10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006244025.2 | XP_006244087.1 | 11 - 12 | 2278 | 79 | |
XM_008766657.1 | XP_008764879.1 | 11 - 12 | 2335 | 79 | ||
XM_243980.7 | XP_243980.4 | 11 - 12 | 3430 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
