Assay Details
Gene Symbol: Tfdp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006243556.2 | XP_006243618.1 | 517 | 92 | ||
XM_008766479.1 | XP_008764701.1 | 3 - 4 | 195 | 92 | ||
XM_008766480.1 | XP_008764702.1 | 3 - 4 | 194 | 92 | ||
XM_008766481.1 | XP_008764703.1 | 3 - 4 | 195 | 92 | ||
XM_008766487.1 | XP_008764709.1 | 4 - 5 | 324 | 92 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008766487.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
