Assay Details
Gene Symbol: Ephb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105868.1 | NP_001099338.1 | 3 - 4 | 1242 | 64 | |
XM_006248561.2 | XP_006248623.1 | 3 - 4 | 1370 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
