Assay Details
Gene Symbol: Tango2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108323.1 | NP_001101793.1 | 3 - 4 | 279 | 77 | |
XM_006248628.2 | XP_006248690.1 | 3 - 4 | 404 | 77 | ||
XM_006248629.2 | XP_006248691.1 | 3 - 4 | 274 | 77 | ||
XM_006248630.2 | XP_006248692.1 | 3 - 4 | 299 | 77 | ||
XM_006248632.2 | XP_006248694.1 | 3 - 4 | 404 | 77 | ||
XM_006248633.2 | XP_006248695.1 | 3 - 4 | 403 | 77 | ||
XM_006248634.2 | XP_006248696.1 | 3 - 4 | 405 | 77 | ||
XM_008768888.1 | XP_008767110.1 | 3 - 4 | 277 | 77 | ||
XM_008768889.1 | XP_008767111.1 | 3 - 4 | 404 | 77 | ||
GenBank mRNA | FQ213583.1 | - | 2 - 3 | 306 | 77 | |
FQ217948.1 | - | 3 - 4 | 406 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
