Assay Details
Gene Symbol: P2rx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012721.2 | NP_036853.2 | 8 - 9 | 1001 | 75 | |
XM_008768851.1 | XP_008767073.1 | 8 - 9 | 1191 | 75 | ||
XM_008768852.1 | XP_008767074.1 | 8 - 9 | 876 | 75 | ||
GenBank mRNA | X92070.1 | - | 8 - 9 | 882 | 75 | |
X97376.1 | - | 8 - 9 | 1012 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
