Assay Details
Gene Symbol: Fam131a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002727913.4 | XP_002727959.1 | 2 - 3 | 480 | 68 | |
XM_008768832.1 | XP_008767054.1 | 3 - 4 | 744 | 68 |
Gene Symbol: LOC100912498 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751082.3 | XP_003751130.1 | 480 | 68 | ||
XM_008768819.1 | XP_008767041.1 | 203 | 68 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008768819.1, XM_008768832.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
