Assay Details
Gene Symbol: Ehhadh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133606.1 | NP_598290.1 | 4 - 5 | 478 | 81 | |
GenBank mRNA | BC089777.1 | - | 4 - 5 | 467 | 81 | |
K03249.1 | - | 4 - 5 | 478 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
