Assay Details
Gene Symbol: Efcab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106930.1 | NP_001100400.1 | 1 - 2 | 179 | 102 | |
GenBank mRNA | FQ216783.1 | - | 1 - 2 | 160 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
