Assay Details
Gene Symbol: Efcab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106930.1 | NP_001100400.1 | 5 - 6 | 698 | 151 | |
XM_008768871.1 | XP_008767093.1 | 5 - 6 | 729 | 151 | ||
GenBank mRNA | FQ216783.1 | - | 4 - 5 | 574 | 151 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
