Assay Details
Gene Symbol: Sept5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053931.3 | NP_446383.3 | 1 - 2 | 333 | 120 | |
GenBank mRNA | AB027143.1 | - | 1 - 2 | 333 | 120 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AB027143.1, NM_053931.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
