Assay Details
Gene Symbol: Klhl24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181473.2 | NP_852138.2 | 6 - 7 | 1750 | 93 | |
GenBank mRNA | AB101615.1 | - | 5 - 6 | 1602 | 93 | |
BC092204.1 | - | 6 - 7 | 1750 | 93 | ||
FQ219171.1 | - | 7 - 8 | 1833 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_43_P21118 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 228577 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 404237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934372 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 125785 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258254 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 30342 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590759 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 268505 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215133 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 337761 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 38267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 547872 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964932 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 826079 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588054 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418957 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682997 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573219 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 609092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 288119 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 694667 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7842 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 154115 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 125257 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P21118 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_43_P21118 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_43_P21118 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P835214 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_43_P21118 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 13396 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 30310 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 102225 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10755538 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10755541 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10755540 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10755539 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10755536 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10755535 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10755534 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10755537 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10755543 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10755533 | - | - |
Rat Genome 230 2.0 Array | 1383110_PM_at | - | - |
Rat Genome 230 2.0 Array | 1386611_PM_at | - | - |
Rat Genome 230 2.0 Array | 1386611_at | - | - |
Rat Genome 230 2.0 Array | 1383110_at | - | - |
Rat Genome 230 2.0 Array | 1383300_at | - | - |
Rat Genome 230 2.0 Array | 1383300_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1383110_at | - | - |
Affymetrix HT Rat Focus Array | 1383300_at | - | - |
Affymetrix HT Rat Focus Array | 1383300_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1383110_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1386611_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1386611_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1100012813.rn.1 | TC1100001954.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1100012815.rn.1 | TC1100001954.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1100012814.rn.1 | TC1100001954.rn.1 | RTA TC |
Rat Expression Set 230 | 1386611_PM_at | - | - |
Rat Expression Set 230 | 1383110_PM_at | - | - |
Rat Expression Set 230 | 1383300_PM_at | - | - |
Rat Expression Set 230 | 1386611_at | - | - |
Rat Expression Set 230 | 1383110_at | - | - |
Rat Expression Set 230 | 1383300_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10755533 | - | - |
Rat Genome U34 Set | rc_AA874906_at | - | - |
Rat Genome U34 Set | rc_AI145238_at | - | - |
Rat Genome U34 Set | rc_AI233404_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21083578 | - | - |
ABI Rat Genome Survey Microarray | 21452899 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1651132 | - | - |
RatRef-12 Expression BeadChip | ILMN_61218 | - | - |
