Assay Details
Gene Symbol: Ydjc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768860.1 | XP_008767082.1 | 2 - 3 | 449 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008768860.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
