Assay Details
Gene Symbol: Ydjc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013863.2 | NP_001013885.2 | 2 - 3 | 332 | 79 | |
XM_006248682.2 | XP_006248744.1 | 2 - 3 | 501 | 79 | ||
XM_008768859.1 | XP_008767081.1 | 3 - 4 | 365 | 79 | ||
XM_008768860.1 | XP_008767082.1 | 4 - 5 | 781 | 79 | ||
XM_008768861.1 | XP_008767083.1 | 4 - 5 | 530 | 79 | ||
GenBank mRNA | BC086565.1 | - | 2 - 3 | 482 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
