Assay Details
Gene Symbol: Ydjc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013863.2 | NP_001013885.2 | 4 - 5 | 616 | 62 | |
XM_006248682.2 | XP_006248744.1 | 4 - 5 | 785 | 62 | ||
XM_008768859.1 | XP_008767081.1 | 5 - 6 | 649 | 62 | ||
XM_008768860.1 | XP_008767082.1 | 6 - 7 | 1065 | 62 | ||
XM_008768861.1 | XP_008767083.1 | 6 - 7 | 814 | 62 | ||
GenBank mRNA | BC086565.1 | - | 4 - 5 | 766 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
