Assay Details
Gene Symbol: Lpp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013864.1 | NP_001013886.1 | 9 - 10 | 1572 | 80 | |
XM_006248505.2 | XP_006248567.1 | 12 - 13 | 2009 | 80 | ||
XM_006248507.2 | XP_006248569.1 | 6 - 7 | 9475 | 80 | ||
GenBank mRNA | BC083627.1 | - | 9 - 10 | 1572 | 7134282 | 80 |
BC083890.1 | - | 8 - 9 | 1510 | 7132481 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
