Assay Details
Gene Symbol: Comt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012531.2 | NP_036663.1 | 3 - 4 | 626 | 111 | |
XM_006248603.2 | XP_006248665.1 | 3 - 4 | 627 | 111 | ||
GenBank mRNA | BC081850.1 | - | 3 - 4 | 484 | 7191956 | 111 |
FQ209401.1 | - | 2 - 3 | 339 | 111 | ||
FQ209488.1 | - | 2 - 3 | 345 | 111 | ||
FQ209638.1 | - | 2 - 3 | 339 | 111 | ||
FQ209670.1 | - | 2 - 3 | 339 | 111 | ||
FQ209706.1 | - | 2 - 3 | 350 | 111 | ||
FQ210267.1 | - | 2 - 3 | 339 | 111 | ||
FQ210416.1 | - | 2 - 3 | 367 | 111 | ||
FQ210430.1 | - | 2 - 3 | 342 | 111 | ||
FQ218190.1 | - | 2 - 3 | 341 | 111 | ||
FQ218375.1 | - | 2 - 3 | 349 | 111 | ||
FQ218564.1 | - | 2 - 3 | 341 | 111 | ||
FQ218595.1 | - | 2 - 3 | 341 | 111 | ||
FQ219257.1 | - | 2 - 3 | 346 | 111 | ||
FQ219433.1 | - | 2 - 3 | 341 | 111 | ||
FQ223536.1 | - | 2 - 3 | 341 | 111 | ||
FQ228829.1 | - | 2 - 3 | 339 | 111 | ||
FQ228870.1 | - | 2 - 3 | 347 | 111 | ||
FQ229523.1 | - | 2 - 3 | 321 | 111 | ||
M60753.1 | - | 430 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
