Assay Details
Gene Symbol: Ahsg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012898.4 | NP_037030.1 | 742 | 62 | ||
GenBank mRNA | BC091118.1 | - | 720 | 62 | ||
FQ209417.1 | - | 742 | 62 | |||
FQ209422.1 | - | 741 | 62 | |||
FQ209502.1 | - | 741 | 62 | |||
FQ209545.1 | - | 741 | 62 | |||
FQ209568.1 | - | 741 | 62 | |||
FQ209654.1 | - | 741 | 62 | |||
FQ209655.1 | - | 738 | 62 | |||
FQ209665.1 | - | 741 | 62 | |||
FQ209680.1 | - | 741 | 62 | |||
FQ209720.1 | - | 741 | 62 | |||
FQ209759.1 | - | 741 | 62 | |||
FQ209858.1 | - | 739 | 62 | |||
FQ209920.1 | - | 741 | 62 | |||
FQ209985.1 | - | 741 | 62 | |||
FQ210056.1 | - | 739 | 62 | |||
FQ210141.1 | - | 741 | 62 | |||
FQ210159.1 | - | 741 | 62 | |||
FQ210164.1 | - | 739 | 62 | |||
FQ210199.1 | - | 741 | 62 | |||
FQ210239.1 | - | 741 | 62 | |||
FQ210249.1 | - | 741 | 62 | |||
FQ210260.1 | - | 741 | 62 | |||
FQ210304.1 | - | 741 | 62 | |||
FQ210382.1 | - | 741 | 62 | |||
FQ210396.1 | - | 741 | 62 | |||
FQ210402.1 | - | 741 | 62 | |||
FQ210405.1 | - | 741 | 62 | |||
FQ210457.1 | - | 741 | 62 | |||
FQ210498.1 | - | 741 | 62 | |||
FQ210504.1 | - | 741 | 62 | |||
FQ210525.1 | - | 740 | 62 | |||
FQ210533.1 | - | 343 | 62 | |||
FQ210731.1 | - | 739 | 62 | |||
FQ210739.1 | - | 741 | 62 | |||
FQ210791.1 | - | 135 | 62 | |||
FQ210802.1 | - | 404 | 62 | |||
FQ210891.1 | - | 741 | 62 | |||
FQ210896.1 | - | 739 | 62 | |||
FQ210953.1 | - | 741 | 62 | |||
FQ210957.1 | - | 747 | 62 | |||
FQ211020.1 | - | 741 | 62 | |||
FQ211045.1 | - | 738 | 62 | |||
FQ211092.1 | - | 741 | 62 | |||
FQ211177.1 | - | 741 | 62 | |||
FQ211299.1 | - | 741 | 62 | |||
FQ211301.1 | - | 742 | 62 | |||
FQ211303.1 | - | 741 | 62 | |||
FQ211304.1 | - | 741 | 62 | |||
FQ211379.1 | - | 741 | 62 | |||
FQ217075.1 | - | 739 | 62 | |||
FQ218089.1 | - | 741 | 62 | |||
FQ218236.1 | - | 741 | 62 | |||
FQ218262.1 | - | 741 | 62 | |||
FQ218269.1 | - | 741 | 61 | |||
FQ218319.1 | - | 340 | 62 | |||
FQ218400.1 | - | 741 | 62 | |||
FQ218410.1 | - | 548 | 62 | |||
FQ218479.1 | - | 741 | 62 | |||
FQ218487.1 | - | 741 | 62 | |||
FQ218489.1 | - | 741 | 62 | |||
FQ218493.1 | - | 741 | 62 | |||
FQ218494.1 | - | 234 | 62 | |||
FQ218502.1 | - | 741 | 62 | |||
FQ218533.1 | - | 741 | 62 | |||
FQ218563.1 | - | 739 | 62 | |||
FQ218569.1 | - | 739 | 62 | |||
FQ218608.1 | - | 741 | 62 | |||
FQ218630.1 | - | 737 | 62 | |||
FQ218687.1 | - | 569 | 62 | |||
FQ218765.1 | - | 741 | 62 | |||
FQ218796.1 | - | 741 | 62 | |||
FQ218858.1 | - | 738 | 62 | |||
FQ218862.1 | - | 741 | 62 | |||
FQ218872.1 | - | 741 | 62 | |||
FQ218877.1 | - | 739 | 62 | |||
FQ218907.1 | - | 741 | 62 | |||
FQ218912.1 | - | 740 | 62 | |||
FQ218931.1 | - | 740 | 62 | |||
FQ218970.1 | - | 739 | 62 | |||
FQ219018.1 | - | 741 | 62 | |||
FQ219028.1 | - | 741 | 62 | |||
FQ219042.1 | - | 740 | 62 | |||
FQ219099.1 | - | 741 | 62 | |||
FQ219129.1 | - | 739 | 62 | |||
FQ219173.1 | - | 741 | 62 | |||
FQ219203.1 | - | 741 | 62 | |||
FQ219221.1 | - | 741 | 62 | |||
FQ219232.1 | - | 747 | 62 | |||
FQ219235.1 | - | 741 | 62 | |||
FQ219295.1 | - | 741 | 62 | |||
FQ219300.1 | - | 741 | 62 | |||
FQ219311.1 | - | 741 | 62 | |||
FQ219326.1 | - | 741 | 62 | |||
FQ219364.1 | - | 741 | 62 | |||
FQ219370.1 | - | 741 | 62 | |||
FQ219400.1 | - | 741 | 62 | |||
FQ219488.1 | - | 741 | 62 | |||
FQ219536.1 | - | 740 | 62 | |||
FQ219546.1 | - | 741 | 62 | |||
FQ219551.1 | - | 741 | 62 | |||
FQ219565.1 | - | 741 | 62 | |||
FQ219587.1 | - | 739 | 62 | |||
FQ219616.1 | - | 476 | 62 | |||
FQ219617.1 | - | 741 | 62 | |||
FQ219643.1 | - | 741 | 62 | |||
FQ219653.1 | - | 740 | 62 | |||
FQ219660.1 | - | 741 | 62 | |||
FQ219663.1 | - | 741 | 62 | |||
FQ219672.1 | - | 741 | 62 | |||
FQ219688.1 | - | 739 | 62 | |||
FQ219755.1 | - | 741 | 62 | |||
FQ219769.1 | - | 741 | 62 | |||
FQ219772.1 | - | 740 | 62 | |||
FQ219773.1 | - | 741 | 62 | |||
FQ219774.1 | - | 741 | 62 | |||
M29758.1 | - | 742 | 62 | |||
X63446.1 | - | 700 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
