Assay Details
Gene Symbol: Trmt2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011895.1 | NP_001011895.1 | 10 - 11 | 1764 | 82 | |
XM_006248604.1 | XP_006248666.1 | 10 - 11 | 1976 | 82 | ||
XM_006248605.1 | XP_006248667.1 | 10 - 11 | 1735 | 82 | ||
GenBank mRNA | BC083619.1 | - | 10 - 11 | 1764 | 7133938 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
