Assay Details
Gene Symbol: Trmt2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011895.1 | NP_001011895.1 | 11 - 12 | 1866 | 61 | |
XM_006248604.1 | XP_006248666.1 | 11 - 12 | 2078 | 61 | ||
XM_006248605.1 | XP_006248667.1 | 11 - 12 | 1837 | 61 | ||
GenBank mRNA | BC083619.1 | - | 11 - 12 | 1866 | 7133938 | 61 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
