Assay Details
Gene Symbol: Tbccd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012016.1 | NP_001012016.1 | 5 - 6 | 1146 | 85 | |
XM_006248545.2 | XP_006248607.1 | 4 - 5 | 1120 | 85 | ||
XM_006248546.2 | XP_006248608.1 | 4 - 5 | 1268 | 85 | ||
GenBank mRNA | BC079273.1 | - | 5 - 6 | 1176 | 85 | |
BC089823.1 | - | 5 - 6 | 1146 | 7133137 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
