Assay Details
Gene Symbol: Smpd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768878.1 | XP_008767100.1 | 12 - 13 | 1355 | 89 | |
GenBank mRNA | FQ211899.1 | - | 12 - 13 | 1390 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
