Assay Details
Gene Symbol: Liph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044279.1 | NP_001037744.1 | 504 | 143 | ||
GenBank mRNA | BC062045.1 | - | 510 | 143 | ||
BC107932.1 | - | 504 | 7124552 | 143 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
