Assay Details
Gene Symbol: Liph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044279.1 | NP_001037744.1 | 611 | 77 | ||
GenBank mRNA | BC062045.1 | - | 617 | 77 | ||
BC107932.1 | - | 611 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
