Assay Details
Gene Symbol: Polr2h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134789.1 | NP_001128261.1 | 1 - 2 | 164 | 135 | |
GenBank mRNA | CB758009.1 | - | 1 - 2 | 177 | 136 | |
CF113682.1 | - | 1 - 2 | 137 | 135 | ||
FQ222889.1 | - | 1 - 2 | 162 | 135 |
Gene Symbol: LOC100912534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751083.2 | XP_003751131.1 | 1 - 2 | 178 | 135 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
