Assay Details
Gene Symbol: Polr2h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001134789.1 | NP_001128261.1 | 4 - 5 | 426 | 56 | |
GenBank mRNA | BQ210721.1 | - | 1 - 2 | 56 | ||
CF113682.1 | - | 4 - 5 | 399 | 56 | ||
FQ222889.1 | - | 4 - 5 | 424 | 56 |
Gene Symbol: LOC100912534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751083.2 | XP_003751131.1 | 4 - 5 | 440 | 56 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
